accessibility menu, dialog, popup

UserName

CAS RN 1667-10-3

ClCl

CAS RN 1667-10-3

IUPAC Name: 4,4'-bis(chloromethyl)-1,1'-biphenyl
Synonyms: 4,4'-bis(chloromethyl)-1,1'-biphenyl
Molecular Weight (g/mol): 251.15
Molecular Formula: C14H12Cl2
InChi Key: INZDTEICWPZYJM-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(C=C1)C1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results