missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(chloromethyl)biphenyl 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 141°C |
| Color | White-Yellow |
| Boiling Point | 184°C |
| Quantity | 500 g |
| UN Number | 3261 |
| Formula Weight | 251.15 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Bis(chloromethyl)biphenyl |
Safety and Handling
| EINECSNumber | (4)-0798 |
| TSCA | No |