missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(4-aminophenoxy)biphenyl 98.0+%, TCI America™
$100.67 - $307.57
Chemical Identifiers
| CAS | 13080-85-8 |
|---|---|
| Molecular Formula | C24H20N2O2 |
| Molecular Weight (g/mol) | 368.44 |
| MDL Number | MFCD00039151 |
| InChI Key | HYDATEKARGDBKU-UHFFFAOYSA-N |
| PubChem CID | 632695 |
| IUPAC Name | 4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline |
| SMILES | NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1 |
Chemical Identifiers
| 13080-85-8 | |
| 368.44 | |
| HYDATEKARGDBKU-UHFFFAOYSA-N | |
| 4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline |
| C24H20N2O2 | |
| MFCD00039151 | |
| 632695 | |
| NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1 |
Specifications
| 13080-85-8 | |
| Gray-White | |
| MFCD00039151 | |
| HYDATEKARGDBKU-UHFFFAOYSA-N | |
| 4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline | |
| 632695 | |
| ≥98.0% (T) | |
| 4,4′-Bis(4-aminophenoxy)biphenyl |
| 199°C | |
| C24H20N2O2 | |
| 5 g | |
| NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1 | |
| 368.44 | |
| 368.44 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-1897
TSCA : No