missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(4-aminophenoxy)biphenyl 98.0+%, TCI America™
Supplier: TCI America B167125G
Specifications
| 4,4′-Bis(4-aminophenoxy)biphenyl | |
| 199°C | |
| C24H20N2O2 | |
| 25 g | |
| NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1 | |
| 368.44 | |
| 368.44 | |
| Crystalline Powder |
| 13080-85-8 | |
| Gray-White | |
| MFCD00039151 | |
| HYDATEKARGDBKU-UHFFFAOYSA-N | |
| 4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline | |
| 632695 | |
| ≥98.0% (T) |
Chemical Identifiers
| 13080-85-8 | |
| 368.44 | |
| HYDATEKARGDBKU-UHFFFAOYSA-N | |
| 4-{[4'-(4-aminophenoxy)-[1,1'-biphenyl]-4-yl]oxy}aniline |
| C24H20N2O2 | |
| MFCD00039151 | |
| 632695 | |
| NC1=CC=C(OC2=CC=C(C=C2)C2=CC=C(OC3=CC=C(N)C=C3)C=C2)C=C1 |
Safety and Handling
EINECSNumber : (4)-1897
TSCA : No