missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldithiol 98.0+%, TCI America™

Catalog No. B22851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22851G Supplier TCI America Supplier No. B22851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6954-27-4
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.332
MDL Number MFCD00046826
InChI Key VRPKUXAKHIINGG-UHFFFAOYSA-N
Synonym 4,4′C-Dimercaptobiphenyl
PubChem CID 81399
ChEBI CHEBI:34363
IUPAC Name 4-(4-sulfanylphenyl)benzenethiol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)S)S

Specifications

Melting Point 183°C
Color Yellow
Quantity 1 g
Formula Weight 218.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldithiol