missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Acetoxyphenyl)-2-butanone 96.0+%, TCI America™

Catalog No. A236825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A236825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A236825G Supplier TCI America Supplier No. A236825G
Only null left

Chemical Identifiers

CAS 3572-06-3
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00008704
InChI Key UMIKWXDGXDJQJK-UHFFFAOYSA-N
Synonym 4-(3-Oxobutyl)phenyl Acetate, Acetic Acid 4-(3-Oxobutyl)phenyl Ester
PubChem CID 19137
IUPAC Name [4-(3-oxobutyl)phenyl] acetate
SMILES CC(=O)CCC1=CC=C(C=C1)OC(=O)C

Specifications

Color Yellow
Boiling Point 124°C
Quantity 25 g
Formula Weight 206.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(4-Acetoxyphenyl)-2-butanone
RTECSNumber EL8950000
TSCA Yes