accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 3572-06-3

IUPAC Name: 4-(3-oxobutyl)phenyl acetate
Synonyms: oxobutylphenyl acetate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: UMIKWXDGXDJQJK-UHFFFAOYSA-N
SMILES: CC(=O)CCC1=CC=C(OC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results