missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 98.0+%, TCI America™
Supplier: TCI America T32611G
Specifications
| 4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane | |
| 179°C | |
| C30H27BO2 | |
| 1 g | |
| NZTBACVVLACPNL-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane | |
| 58489416 | |
| ≥98.0% (GC,T) |
| 1115639-92-3 | |
| White-Yellow | |
| MFCD28130383 | |
| 2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene | |
| CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1 | |
| 430.35 | |
| 430.35 | |
| Crystalline Powder |
Chemical Identifiers
| 1115639-92-3 | |
| 430.35 | |
| NZTBACVVLACPNL-UHFFFAOYSA-N | |
| 58489416 | |
| CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1 |
| C30H27BO2 | |
| MFCD28130383 | |
| 2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene | |
| 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No