missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 98.0+%, TCI America™
$65.24 - $193.86
Chemical Identifiers
| CAS | 1115639-92-3 |
|---|---|
| Molecular Formula | C30H27BO2 |
| Molecular Weight (g/mol) | 430.35 |
| MDL Number | MFCD28130383 |
| InChI Key | NZTBACVVLACPNL-UHFFFAOYSA-N |
| Synonym | 2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene |
| PubChem CID | 58489416 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane |
| SMILES | CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1 |
Chemical Identifiers
| 1115639-92-3 | |
| 430.35 | |
| NZTBACVVLACPNL-UHFFFAOYSA-N | |
| 58489416 | |
| CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1 |
| C30H27BO2 | |
| MFCD28130383 | |
| 2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene | |
| 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane |
Specifications
| 1115639-92-3 | |
| White-Yellow | |
| MFCD28130383 | |
| 2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene | |
| CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1 | |
| 430.35 | |
| 430.35 | |
| Crystalline Powder |
| 179°C | |
| C30H27BO2 | |
| 200 mg | |
| NZTBACVVLACPNL-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane | |
| 58489416 | |
| ≥98.0% (GC,T) | |
| 4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No