missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane 97.0+%, TCI America™
Supplier: TCI America T30891G
Specifications
| 4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane | |
| 237°C | |
| 1 g | |
| RKJWQQVQQVALBZ-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-perylen-3-yl-1,3,2-dioxaborolane | |
| 24751681 | |
| ≥97.0% (GC) |
| 950761-81-6 | |
| C26H23BO2 | |
| (3-Perylenyl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)perylene | |
| B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=C4C3=C(C=C2)C5=CC=CC6=C5C4=CC=C6 | |
| 378.278 | |
| 378.28 | |
| Crystalline Powder |
Chemical Identifiers
| 950761-81-6 | |
| 378.278 | |
| (3-Perylenyl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)perylene | |
| 4,4,5,5-tetramethyl-2-perylen-3-yl-1,3,2-dioxaborolane |
| C26H23BO2 | |
| RKJWQQVQQVALBZ-UHFFFAOYSA-N | |
| 24751681 | |
| B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=C4C3=C(C=C2)C5=CC=CC6=C5C4=CC=C6 |
Safety and Handling
TSCA : No