missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(3,4,5-trifluorophenyl)-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. T33761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33761G 1 g
T33765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33761G Supplier TCI America Supplier No. T33761G

Chemical Identifiers

CAS 827614-70-0
Molecular Formula C12H14BF3O2
Molecular Weight (g/mol) 258.047
MDL Number MFCD05663885
InChI Key VFCTUUBAONBDJU-UHFFFAOYSA-N
Synonym 3,4,5-Trifluorophenylboronic Acid Pinacol Ester, 1,2,3-Trifluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
PubChem CID 2760701
IUPAC Name 4,4,5,5-tetramethyl-2-(3,4,5-trifluorophenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 258.05
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(3,4,5-trifluorophenyl)-1,3,2-dioxaborolane