missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™

Catalog No. p-7137039
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19541G 1 g
T19545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19541G Supplier TCI America Supplier No. T19541G

Chemical Identifiers

CAS 269409-70-3
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.075
MDL Number MFCD02093756
InChI Key BICZJRAGTCRORZ-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
PubChem CID 2734624
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O

Specifications

Melting Point 118°C
Quantity 1 g
Formula Weight 220.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol