missing translation for 'onlineSavingsMsg'
Learn More

4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide 98.0+%, TCI America™

Catalog No. T19501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19501G Supplier TCI America Supplier No. T19501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 214360-60-8
Molecular Formula C14H20BNO3
Molecular Weight (g/mol) 261.13
MDL Number MFCD02093722
InChI Key ANGKVUVZQVUVJO-UHFFFAOYSA-N
Synonym n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetamide, 4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl acetanilide, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl acetanilide, 4-acetamidophenylboronic acid pinacol ester, n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetamide, acetamide, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, 4-acetamidophenylboronic acid, pinacol ester, 4-acetylaminophenylboronic acid pinacol ester, 2-4-acetamidophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2734619
IUPAC Name N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide