accessibility menu, dialog, popup

UserName

CH3OHNBOH3CH3CH3CCH3O

CAS RN 214360-60-8

IUPAC Name: N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide
Synonyms: N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide
Molecular Weight (g/mol): 261.13
Molecular Formula: C14H20BNO3
InChi Key: ANGKVUVZQVUVJO-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results