missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic Anhydride 98.0+%, TCI America™

Catalog No. i085625g
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
I085625G 25 g
I0856100G 100 g
2 options

Catalog No. I085625G

Supplier: TCI America I085625G

Only null left

Chemical Identifiers

CAS 38103-06-9
Molecular Formula C31H20O8
Molecular Weight (g/mol) 520.49
MDL Number MFCD00319167
InChI Key MQAHXEQUBNDFGI-UHFFFAOYSA-N
PubChem CID 94483
IUPAC Name 5-[4-(2-{4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]phenyl}propan-2-yl)phenoxy]-1,3-dihydro-2-benzofuran-1,3-dione
SMILES CC(C)(C1=CC=C(OC2=CC=C3C(=O)OC(=O)C3=C2)C=C1)C1=CC=C(OC2=CC=C3C(=O)OC(=O)C3=C2)C=C1

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 25 g
Synonym 4,4′C-(4,4′C-Isopropylidenediphenoxy)bis(phthalic Anhydride)
Formula Weight 520.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-(4,4′-Isopropylidenediphenoxy)diphthalic Anhydride
RTECSNumber NP7227500
TSCA Yes