accessibility menu, dialog, popup

UserName

CAS RN 38103-06-9

H3CCH3OOOOOOOO

CAS RN 38103-06-9

IUPAC Name: 5-[4-(2-{4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]phenyl}propan-2-yl)phenoxy]-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-[4-(2-{4-[(1,3-dioxo-2-benzofuran-5-yl)oxy]phenyl}propan-2-yl)phenoxy]-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 520.49
Molecular Formula: C31H20O8
InChi Key: MQAHXEQUBNDFGI-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(OC2=CC=C3C(=O)OC(=O)C3=C2)C=C1)C1=CC=C(OC2=CC=C3C(=O)OC(=O)C3=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results