missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Hydroxyphenoxy)benzoic Acid 99.0+%, TCI America™

Catalog No. H06415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H06415G Supplier TCI America Supplier No. H06415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56183-35-8
Molecular Formula C13H10O4
Molecular Weight (g/mol) 230.219
MDL Number MFCD00059644
InChI Key DEDKCEDXZJIDKZ-UHFFFAOYSA-N
PubChem CID 615389
IUPAC Name 4-(3-hydroxyphenoxy)benzoic acid
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)C(=O)O)O

Specifications

Melting Point 171°C
Color Yellow
Quantity 5 g
Formula Weight 230.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Hydroxyphenoxy)benzoic Acid