accessibility menu, dialog, popup

UserName

OOOHHO

CAS RN 56183-35-8

IUPAC Name: 4-(3-hydroxyphenoxy)benzoic acid
Synonyms: 4-(3-hydroxyphenoxy)benzoic acid
Molecular Weight (g/mol): 230.22
Molecular Formula: C13H10O4
InChi Key: DEDKCEDXZJIDKZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=CC(O)=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results