missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Butenyloxy)benzoic Acid 98.0+%, TCI America™

Catalog No. B15855G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15855G Supplier TCI America Supplier No. B15855G

Chemical Identifiers

CAS 115595-27-2
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00191325
InChI Key QXZIOUAINSTHGI-UHFFFAOYSA-N
PubChem CID 544452
IUPAC Name 4-(but-3-en-1-yloxy)benzoic acid
SMILES OC(=O)C1=CC=C(OCCC=C)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Butenyloxy)benzoic Acid