accessibility menu, dialog, popup

UserName

CAS RN 115595-27-2

H2COOOH

CAS RN 115595-27-2

IUPAC Name: 4-(but-3-en-1-yloxy)benzoic acid
Synonyms: 4-(but-3-en-1-yloxy)benzoic acid
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: QXZIOUAINSTHGI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OCCC=C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results