missing translation for 'onlineSavingsMsg'
Learn More

4-(3,4-Dichlorophenyl)-1-tetralone 98.0+%, TCI America™

Catalog No. p-7128450
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47661G 1 g
D47665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47661G Supplier TCI America Supplier No. D47661G

Chemical Identifiers

CAS 79560-19-3
Molecular Formula C16H12Cl2O
Molecular Weight (g/mol) 291.171
MDL Number MFCD02093088
InChI Key JGMBHJNMQVKDMW-UHFFFAOYSA-N
Synonym 4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
PubChem CID 3746251
IUPAC Name 4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC(=O)C2=CC=CC=C2C1C3=CC(=C(C=C3)Cl)Cl

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Formula Weight 291.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3,4-Dichlorophenyl)-1-tetralone