CAS RN 79560-19-3
IUPAC Name:
4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms:
4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol):
291.17
Molecular Formula:
C16H12Cl2O
InChi Key:
JGMBHJNMQVKDMW-UHFFFAOYNA-N
SMILES:
ClC1=C(Cl)C=C(C=C1)C1CCC(=O)C2=CC=CC=C12
1
–
1
of
1
results
1
4-(3,4-Dichlorophenyl)-1-tetralone 98.0+%, TCI America™
CAS: 79560-19-3 Molecular Formula: C16H12Cl2O Molecular Weight (g/mol): 291.171 MDL Number: MFCD02093088 InChI Key: JGMBHJNMQVKDMW-UHFFFAOYSA-N Synonym: 4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone PubChem CID: 3746251 IUPAC Name: 4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one SMILES: C1CC(=O)C2=CC=CC=C2C1C3=CC(=C(C=C3)Cl)Cl