missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)benzoic acid, 96%

Catalog No. AAH3364203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3364203 1 g
AAH3364206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3364203 Supplier Thermo Scientific Chemicals Supplier No. H3364203

CAS: 29886-62-2 | C11H8O2S | 204.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29886-62-2
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
MDL Number MFCD00690742
InChI Key CVDUBQJEQNRCIZ-UHFFFAOYSA-N
Synonym 4-thiophen-2-yl benzoic acid, 4-2-thienyl benzoic acid, 4-thiophen-2-yl-benzoic acid, benzoic acid,4-2-thienyl, benzoic acid, 4-2-thienyl, pubchem12353, acmc-1cb4z, 4-2-thienyl benzoicacid, 4-thien-2-ylbenzoic acid, amtda058
PubChem CID 736498
IUPAC Name 4-thiophen-2-ylbenzoic acid
SMILES C1=CSC(=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 243°C to 245°C
Quantity 1 g
Formula Weight 204.25
Percent Purity 96%
Chemical Name or Material 4-(2-Thienyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only