accessibility menu, dialog, popup

UserName

OOS

CAS RN 29886-62-2

IUPAC Name: 4-(thiophen-2-yl)benzoate
Synonyms: 4-(thiophen-2-yl)benzoate
Molecular Weight (g/mol): 203.24
Molecular Formula: C11H7O2S
InChi Key: CVDUBQJEQNRCIZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results