Learn More
4-(2-Pyrrolidinyl)pyridine, 96%
CAS: 128562-25-4 | C9H12N2 | 148.209 g/mol
Supplier: Thermo Scientific Chemicals H2605103
Description
4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Specifications
| 4-(2-Pyrrolidinyl)pyridine | |
| C9H12N2 | |
| 1 g | |
| Slightly soluble in water. | |
| C1CC(NC1)C2=CC=NC=C2 | |
| 148.209 | |
| 148.21 |
| 128562-25-4 | |
| MFCD01862536 | |
| 4-pyrrolidin-2-yl pyridine, 4-2-pyrrolidinyl pyridine, 4-pyrrolidin-2-yl-pyridine, 2-4-pyridyl pyrrolidine, pyridine, 4-2-pyrrolidinyl, 2-4-pyridinyl pyrrolidine, +/--4-pyrrolidin-2-yl pyridine, 4-2-pyridyl pyrrolidine, 2-pyridin-4-yl pyrrolidine | |
| GDGNPIOGJLCICG-UHFFFAOYSA-N | |
| 4-pyrrolidin-2-ylpyridine | |
| 2771664 | |
| 96% |
Chemical Identifiers
| 128562-25-4 | |
| 148.209 | |
| GDGNPIOGJLCICG-UHFFFAOYSA-N | |
| 2771664 | |
| C1CC(NC1)C2=CC=NC=C2 |
| C9H12N2 | |
| MFCD01862536 | |
| 4-pyrrolidin-2-yl pyridine, 4-2-pyrrolidinyl pyridine, 4-pyrrolidin-2-yl-pyridine, 2-4-pyridyl pyrrolidine, pyridine, 4-2-pyrrolidinyl, 2-4-pyridinyl pyrrolidine, +/--4-pyrrolidin-2-yl pyridine, 4-2-pyridyl pyrrolidine, 2-pyridin-4-yl pyrrolidine | |
| 4-pyrrolidin-2-ylpyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only