missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyrrolidinyl)pyridine, 96%

Catalog No. p-7028652
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26051MD 250 mg
AAH2605103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26051MD Supplier Thermo Scientific Chemicals Supplier No. H26051MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128562-25-4 | C9H12N2 | 148.209 g/mol

4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Pyrrolidinyl)pyridine can be used in aldol catalysis, in the preparation of azole compounds as 11β-HSD1 inhibitors as well as in the asymetric synthesis of pyrazolodihydropyrimidinylpyrrolidin amide derivatives.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.

Chemical Identifiers

CAS 128562-25-4
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD01862536
InChI Key GDGNPIOGJLCICG-UHFFFAOYSA-N
Synonym 4-pyrrolidin-2-yl pyridine, 4-2-pyrrolidinyl pyridine, 4-pyrrolidin-2-yl-pyridine, 2-4-pyridyl pyrrolidine, pyridine, 4-2-pyrrolidinyl, 2-4-pyridinyl pyrrolidine, +/--4-pyrrolidin-2-yl pyridine, 4-2-pyridyl pyrrolidine, 2-pyridin-4-yl pyrrolidine
PubChem CID 2771664
IUPAC Name 4-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CC=NC=C2

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 148.21
Percent Purity 96%
Chemical Name or Material 4-(2-Pyrrolidinyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only