CAS RN 128562-25-4
IUPAC Name:
2-oxo-3-(thiophen-2-yl)propanoate
Synonyms:
2-oxo-3-(thiophen-2-yl)propanoate
Molecular Weight (g/mol):
169.17
Molecular Formula:
C7H5O3S
InChi Key:
DFEKJNVSDUKUIR-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C(=O)CC1=CC=CS1
1
–
1
of
1
results
1
Promotions Available
4-(2-Pyrrolidinyl)pyridine, 96%
4-(2-Pyrrolidinyl)pyridine, 96%
CAS: 128562-25-4 Molecular Formula: C9H12N2 Molecular Weight (g/mol): 148.209 MDL Number: MFCD01862536 InChI Key: GDGNPIOGJLCICG-UHFFFAOYSA-N Synonym: 4-pyrrolidin-2-yl pyridine,4-2-pyrrolidinyl pyridine,4-pyrrolidin-2-yl-pyridine,2-4-pyridyl pyrrolidine,pyridine, 4-2-pyrrolidinyl,2-4-pyridinyl pyrrolidine,+/--4-pyrrolidin-2-yl pyridine,4-2-pyridyl pyrrolidine,2-pyridin-4-yl pyrrolidine PubChem CID: 2771664 IUPAC Name: 4-pyrrolidin-2-ylpyridine SMILES: C1CC(NC1)C2=CC=NC=C2