missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-yl)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7135586
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24991G 1 g
P24995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24991G Supplier TCI America Supplier No. P24991G

Chemical Identifiers

CAS 16209-00-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD02682056
InChI Key XOEKYPIBVOGCDG-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl benzoic acid, 4-1-pyrazolyl benzoic acid, benzoic acid, 4-1h-pyrazol-1-yl, 4-pyrazol-1-yl benzoic acid, 4-pyrazol-1-yl-benzoic acid, 4-pyrazolylbenzoic acid, pubchem19583, acmc-1bo47, benzoic acid, 4-pyrazol-1-yl, 4-1h-pyrazole-1-yl benzoic acid
PubChem CID 736527
IUPAC Name 4-pyrazol-1-ylbenzoic acid
SMILES C1=CN(N=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 269°C
Quantity 1 g
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzoic Acid