accessibility menu, dialog, popup

UserName

OONN

CAS RN 16209-00-0

IUPAC Name: 4-(1H-pyrazol-1-yl)benzoate
Synonyms: 4-(pyrazol-1-yl)benzoate
Molecular Weight (g/mol): 187.18
Molecular Formula: C10H7N2O2
InChi Key: XOEKYPIBVOGCDG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)N1C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results