missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Propynyl)phthalic Anhydride 98.0+%, TCI America™

Supplier:  TCI America M275725G

 View more versions of this product

Catalog No. M275725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(1-Propynyl)phthalic Anhydride
115°C
MFCD28365741
4-(Methylethynyl)phthalic Anhydride
CC#CC1=CC2=C(C=C1)C(=O)OC2=O
186.17
186.17
Crystalline Powder
1240685-26-0
C11H6O3
25 g
LBTIDDCTPLEEAG-UHFFFAOYSA-N
5-(prop-1-yn-1-yl)-1,3-dihydro-2-benzofuran-1,3-dione
21339433
≥98.0% (GC,T)

Chemical Identifiers

1240685-26-0
186.17
LBTIDDCTPLEEAG-UHFFFAOYSA-N
21339433
CC#CC1=CC2=C(C=C1)C(=O)OC2=O
C11H6O3
MFCD28365741
4-(Methylethynyl)phthalic Anhydride
5-(prop-1-yn-1-yl)-1,3-dihydro-2-benzofuran-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions