missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Propynyl)phthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7133410
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27575G Supplier TCI America Supplier No. M27575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1240685-26-0
Molecular Formula C11H6O3
Molecular Weight (g/mol) 186.17
MDL Number MFCD28365741
InChI Key LBTIDDCTPLEEAG-UHFFFAOYSA-N
Synonym 4-(Methylethynyl)phthalic Anhydride
PubChem CID 21339433
IUPAC Name 5-(prop-1-yn-1-yl)-1,3-dihydro-2-benzofuran-1,3-dione
SMILES CC#CC1=CC2=C(C=C1)C(=O)OC2=O

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 186.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Propynyl)phthalic Anhydride