accessibility menu, dialog, popup

UserName

CAS RN 1240685-26-0

IUPAC Name: 5-(prop-1-yn-1-yl)-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-(prop-1-yn-1-yl)-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 186.17
Molecular Formula: C11H6O3
InChi Key: LBTIDDCTPLEEAG-UHFFFAOYSA-N
SMILES: CC#CC1=CC2=C(C=C1)C(=O)OC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results