missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Piperazinyl)phenol, 95%

Catalog No. p-7024876
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2012218 50 g
AAB2012230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2012218 Supplier Thermo Scientific Chemicals Supplier No. B2012218

CAS: 56621-48-8 | C10H14N2O | 178.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56621-48-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD00066156
InChI Key GPEOAEVZTOQXLG-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl piperazine, 4-piperazin-1-yl phenol, 4-1-piperazinyl phenol, n-4-hydroxyphenyl piperazine, p-1-piperazinyl phenol, 4-piperazinophenol, phenol, 4-1-piperazinyl, 1-4-hydroxyphenyl-piperazine, 4-piperazinylphenol, 4-piperazinyl phenol
PubChem CID 92467
SMILES OC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 218°C to 224°C
Quantity 50 g
UN Number UN3263
Beilstein 150964
Sensitivity Air Sensitive
IUPAC Name 4-(piperazin-1-yl)phenol
Formula Weight 178.24
Percent Purity 98%
Chemical Name or Material 4-(1-Piperazinyl)phenol
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 260-289-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only