accessibility menu, dialog, popup

UserName

CAS RN 56621-48-8

IUPAC Name: 4-(piperazin-1-yl)phenol
Synonyms: 4-(piperazin-1-yl)phenol
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: GPEOAEVZTOQXLG-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N1CCNCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)