missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester, 96%

CAS: 917397-94-5 | C16H19BFNO2 | 287.14 g/mol

Supplier:  Thermo Scientific Chemicals H59075MD

 View more versions of this product

Catalog No. AAH59075MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester
131°C to 133°C
MFCD22418790
4-1-cyanocyclopropyl-2-fluorobenzeneboronic acid pinacol ester, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropane-1-carbonitrile, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropanecarbonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C1(CC1)C#N
287.14
287.14
917397-94-5
C16H19BFNO2
250 mg
TZNIMHCBELUQIQ-UHFFFAOYSA-N
1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbonitrile
69074412
96%

Chemical Identifiers

917397-94-5
287.14
TZNIMHCBELUQIQ-UHFFFAOYSA-N
69074412
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C1(CC1)C#N
C16H19BFNO2
MFCD22418790
4-1-cyanocyclopropyl-2-fluorobenzeneboronic acid pinacol ester, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropane-1-carbonitrile, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropanecarbonitrile
1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only