accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNF

CAS RN 917397-94-5

IUPAC Name: 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbonitrile
Synonyms: 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbonitrile
Molecular Weight (g/mol): 287.14
Molecular Formula: C16H19BFNO2
InChi Key: TZNIMHCBELUQIQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C1(CC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results