missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. p-7039704
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59075MD 250 mg
AAH5907503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59075MD Supplier Thermo Scientific Chemicals Supplier No. H59075MD
Only null left

CAS: 917397-94-5 | C16H19BFNO2 | 287.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 917397-94-5
Molecular Formula C16H19BFNO2
Molecular Weight (g/mol) 287.14
MDL Number MFCD22418790
InChI Key TZNIMHCBELUQIQ-UHFFFAOYSA-N
Synonym 4-1-cyanocyclopropyl-2-fluorobenzeneboronic acid pinacol ester, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropane-1-carbonitrile, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropanecarbonitrile
PubChem CID 69074412
IUPAC Name 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbonitrile
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C1(CC1)C#N

Specifications

Melting Point 131°C to 133°C
Quantity 250 mg
Formula Weight 287.14
Percent Purity 96%
Chemical Name or Material 4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only