missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Bromoethyl)benzoic acid, 98%

Catalog No. p-7046072
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1302503 1 g
AAL1302506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1302503 Supplier Thermo Scientific Chemicals Supplier No. L1302503

CAS: 113023-73-7 | C9H9BrO2 | 229.073 g/mol

4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 113023-73-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00020378
InChI Key VICCYULHZWEWMB-UHFFFAOYSA-N
Synonym 4-1-bromoethyl benzoic acid, benzoic acid, 4-1-bromoethyl, acmc-1c3tf, 4-1-bromoethyl-benzoic acid, 4-1-bromo-ethyl-benzoic acid, benzoic acid,4-1-bromoethyl
PubChem CID 5146225
IUPAC Name 4-(1-bromoethyl)benzoic acid
SMILES CC(C1=CC=C(C=C1)C(=O)O)Br

Specifications

Melting Point 155°C to 156°C
Quantity 1 g
UN Number UN3261
Beilstein 2084039
Solubility Information Insoluble in water.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 4-(1-Bromoethyl)benzoic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only