accessibility menu, dialog, popup

UserName

CAS RN 113023-73-7

IUPAC Name: 4-(1-bromoethyl)benzoic acid
Synonyms: 4-(1-bromoethyl)benzoic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: VICCYULHZWEWMB-UHFFFAOYNA-N
SMILES: CC(Br)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results