missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Dithiolan-2-yl)phenol, 97%

Catalog No. p-7023475
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1706606 5 g
AAA1706614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1706606 Supplier Thermo Scientific Chemicals Supplier No. A1706606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22068-49-1 | C9H10OS2 | 198.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22068-49-1
Molecular Formula C9H10OS2
Molecular Weight (g/mol) 198.30
MDL Number MFCD00068127
InChI Key LTNPCGWCUVDEEY-UHFFFAOYSA-N
Synonym 4-1,3-dithiolan-2-yl phenol, unii-pip8o1r36t, pip8o1r36t, p-1,3-dithiolan-2-yl phenol, phenol, p-1,3-dithiolan-2-yl, 2-4-hydroxyphenyl-1,3-dithiolane, 2-4'-hydroxyphenyl-1,3-dithiolane, 1,3-dithiolane, 2-4-hydroxyphenyl, acmc-1cmgs, phenol,4-1,3-dithiolan-2-yl
PubChem CID 97562
IUPAC Name 4-(1,3-dithiolan-2-yl)phenol
SMILES OC1=CC=C(C=C1)C1SCCS1

Specifications

Melting Point 117°C to 119°C
Quantity 5 g
Formula Weight 198.3
Percent Purity 97%
Chemical Name or Material 4-(1,3-Dithiolan-2-yl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only