CAS RN 22068-49-1
IUPAC Name:
4-(1,3-dithiolan-2-yl)phenol
Synonyms:
4-(1,3-dithiolan-2-yl)phenol
Molecular Weight (g/mol):
198.3
Molecular Formula:
C9H10OS2
InChi Key:
LTNPCGWCUVDEEY-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)C1SCCS1
1
–
1
of
1
results
1
Promotions Available
4-(1,3-Dithiolan-2-yl)phenol, 97%
4-(1,3-Dithiolan-2-yl)phenol, 97%
CAS: 22068-49-1 Molecular Formula: C9H10OS2 Molecular Weight (g/mol): 198.30 MDL Number: MFCD00068127 InChI Key: LTNPCGWCUVDEEY-UHFFFAOYSA-N Synonym: 4-1,3-dithiolan-2-yl phenol,unii-pip8o1r36t,pip8o1r36t,p-1,3-dithiolan-2-yl phenol,phenol, p-1,3-dithiolan-2-yl,2-4-hydroxyphenyl-1,3-dithiolane,2-4'-hydroxyphenyl-1,3-dithiolane,1,3-dithiolane, 2-4-hydroxyphenyl,acmc-1cmgs,phenol,4-1,3-dithiolan-2-yl PubChem CID: 97562 IUPAC Name: 4-(1,3-dithiolan-2-yl)phenol SMILES: OC1=CC=C(C=C1)C1SCCS1