missing translation for 'onlineSavingsMsg'
Learn More

4-(1,1,3,3-Tetramethylbutyl)phenol 95.0+%, TCI America™

Catalog No. T014425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T014425G 25 g
T0144500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T014425G Supplier TCI America Supplier No. T014425G
Only null left

Chemical Identifiers

CAS 140-66-9
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.329
MDL Number MFCD00002368
InChI Key ISAVYTVYFVQUDY-UHFFFAOYSA-N
Synonym 4-tert-octylphenol, p-tert-octylphenol, 4-t-octylphenol, 4-1,1,3,3-tetramethylbutyl phenol, 4-2,4,4-trimethylpentan-2-yl phenol, p-terc.oktylfenol, para-tert-octylphenol, p-1,1,3,3-tetramethylbutyl phenol, phenol, 4-1,1,3,3-tetramethylbutyl, phenol, p-tert-octyl
PubChem CID 8814
ChEBI CHEBI:34445
IUPAC Name 4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)O

Specifications

Melting Point 83°C
Color White
Boiling Point 279°C
Quantity 25 g
UN Number 3077
Formula Weight 206.33
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1,1,3,3-Tetramethylbutyl)phenol
EINECSNumber (3)-0503
RTECSNumber SM9625000
TSCA Yes