accessibility menu, dialog, popup

UserName

H3CH3CCH3H3CCH3OH

CAS RN 140-66-9

IUPAC Name: 4-(2,4,4-trimethylpentan-2-yl)phenol
Synonyms: 4-tert-octylphenol p-terc.oktylfenol
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: ISAVYTVYFVQUDY-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results