missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™
$199.36 - $816.41
Chemical Identifiers
| CAS | 122536-76-9 |
|---|---|
| Molecular Formula | C9H18N2O2 |
| Molecular Weight (g/mol) | 186.255 |
| MDL Number | MFCD00143194 |
| InChI Key | DQQJBEAXSOOCPG-ZETCQYMHSA-N |
| Synonym | s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine |
| PubChem CID | 1514396 |
| IUPAC Name | tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCNC1 |
Chemical Identifiers
| 122536-76-9 | |
| 186.255 | |
| DQQJBEAXSOOCPG-ZETCQYMHSA-N | |
| 1514396 | |
| CC(C)(C)OC(=O)NC1CCNC1 |
| C9H18N2O2 | |
| MFCD00143194 | |
| s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine | |
| tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate |
Specifications
| 78°C | |
| C9H18N2O2 | |
| s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine | |
| CC(C)(C)OC(=O)NC1CCNC1 | |
| 186.255 | |
| 186.26 | |
| Crystalline Powder |
| 122536-76-9 | |
| MFCD00143194 | |
| DQQJBEAXSOOCPG-ZETCQYMHSA-N | |
| tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate | |
| 1514396 | |
| ≥98.0% (GC,T) | |
| (3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine |
Safety and Handling
TSCA : No