missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™

Supplier:  TCI America A117225G

Quantity 25 g

 View more versions of this product

Catalog No. A117225G


Only null left

Description

Description

Specifications

Chemical Identifiers

122536-76-9
186.255
DQQJBEAXSOOCPG-ZETCQYMHSA-N
1514396
CC(C)(C)OC(=O)NC1CCNC1
C9H18N2O2
MFCD00143194
s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate

Specifications

78°C
122536-76-9
MFCD00143194
DQQJBEAXSOOCPG-ZETCQYMHSA-N
tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
186.255
186.26
Crystalline Powder
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
C9H18N2O2
s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
CC(C)(C)OC(=O)NC1CCNC1
25 g
1514396
≥98.0% (GC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions