missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| UN Number | 2735 |
| Formula Weight | 190.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine |
Safety and Handling
| TSCA | No |