accessibility menu, dialog, popup

UserName

CAS RN 169749-99-9

CH3HNN

CAS RN 169749-99-9

IUPAC Name: 1-benzyl-N-methylpyrrolidin-3-amine
Synonyms: 1-benzyl-N-methylpyrrolidin-3-amine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: UEAYAIWNQQWSBK-UHFFFAOYNA-N
SMILES: CNC1CCN(CC2=CC=CC=C2)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results