missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. B15835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15835G 5 g
B158325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15835G Supplier TCI America Supplier No. B15835G

Chemical Identifiers

CAS 169749-99-9
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00191310
InChI Key UEAYAIWNQQWSBK-LBPRGKRZSA-N
PubChem CID 10397666
IUPAC Name (3S)-1-benzyl-N-methylpyrrolidin-3-amine
SMILES CNC1CCN(C1)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 190.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (3S)-(+)-1-Benzyl-3-(methylamino)pyrrolidine