missing translation for 'onlineSavingsMsg'
Learn More

(3R,4R)-1-Benzyl-4-hydroxy-3-pyrrolidinemethanol 96.0+%, TCI America™

Catalog No. H1200200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1200200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1200200MG Supplier TCI America Supplier No. H1200200MG
Only null left

Chemical Identifiers

CAS 253129-03-2
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
InChI Key XQYUBSUGHRDQSN-NEPJUHHUSA-N
PubChem CID 11579361
IUPAC Name (3R,4R)-1-benzyl-4-(hydroxymethyl)pyrrolidin-3-ol
SMILES C1C(C(CN1CC2=CC=CC=C2)O)CO

Specifications

Melting Point 70°C
Quantity 200 mg
Formula Weight 207.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3R,4R)-1-Benzyl-4-hydroxy-3-pyrrolidinemethanol