CAS RN 253129-03-2
IUPAC Name:
1-benzyl-4-(hydroxymethyl)pyrrolidin-3-ol
Synonyms:
1-benzyl-4-(hydroxymethyl)pyrrolidin-3-ol
Molecular Weight (g/mol):
207.27
Molecular Formula:
C12H17NO2
InChi Key:
XQYUBSUGHRDQSN-UHFFFAOYNA-N
SMILES:
OCC1CN(CC2=CC=CC=C2)CC1O
1
–
1
of
1
results
1
(3R,4R)-1-Benzyl-4-hydroxy-3-pyrrolidinemethanol 96.0+%, TCI America™
CAS: 253129-03-2 Molecular Formula: C12H17NO2 Molecular Weight (g/mol): 207.273 InChI Key: XQYUBSUGHRDQSN-NEPJUHHUSA-N PubChem CID: 11579361 IUPAC Name: (3R,4R)-1-benzyl-4-(hydroxymethyl)pyrrolidin-3-ol SMILES: C1C(C(CN1CC2=CC=CC=C2)O)CO